Enclosed Case Carts, Lakeside®
Fournisseur: Lakeside
Certificats
A propos de cet article
Stainless steel carts features fully welded construction, exterior push handles for better control, and more interior space
Positive door latches and sealed doors safeguard contents against cross-contamination. Corner bumpers protect walls and doorways. One piece shell/top minimizes seams and discouraging bacteria growth. Cross-broke floor is angled to the middle with small holes allowing drainage. Units include one pull-out, adjustable-height shelf in the single compartment
Certifications: Built to Sterile Guard specifications.
Ordering information: Additional accessory pull-out and fixed shelves are available; contact your Avantor representative for more information.
Customers Who Bought This Also Bought