Corning® Gosselin™ Straight Containers, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Made of PP, natural or colored.
- Available in sterile or under aseptic conditions packed versions
- Very good chemical and temperature resistance
- Graduated
- Without cap
Ideal for sampling, storage and transport of liquid and solid samples.
Sized to provide adequate volume for milk sampling. It is shock resistant and is assured sterile 10-3.
Corning® Gosselin™ is a Corning brand of laboratory plasticware designed for use in industrial microbiology labs such as Food and Beverage, Environmental/Water Testing, etc.
Customers Who Bought This Also Bought