ContactUsLinkComponent
Howard Mold Counting Chamber, Hausser Scientific
Howard Mold Counting Chamber, Hausser Scientific
# de catalogue 23649-006
Fournisseur:  Hausser
Howard Mold Counting Chamber, Hausser Scientific
# de catalogue 23649-006
Fournisseur:  Hausser
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Howard Mold Counting Chamber
  • Cat. no.:
    23649-006
  • Supplier No.:
    3820
Spécifications
A propos de cet article
The chamber is designed for use in determining mold counts in tomato products
The counting chamber is constructed entirely of glass. The center of the glass slide contains a 15x20mm rectangle that is flanked by 0.1mm shoulders on each side. These shoulders support a cover glass and provide a depth of 0.1mm between the underside of the cover glass and the rectangle. Both the rectangle and the cover glass have optically plane surfaces. To facilitate calibration of the microscope, the rectangle features two engraved, parallel lines spaced 1.382mm apart. The optional accessory eyepiece micrometer is ruled into squares, each of which is equal to 1/6 of the diameter of the eyepiece diaphragm opening. For mold counting, the microscope must give an area of 1.5mm2 (1.382mm in diameter for circle) at a magnification of 90–125.
It may be used with a variety of samples, including canned tomatoes, tomato soup, dehydrated tomatoes, canned spaghetti, or similar products containing tomato sauce.
Ordering information: Counting chamber is supplied with 2 cover slips (72370-012 and 23649-028). These cover slips are available separately. The eyepiece mircometer is not supplied with the counting chamber, and must be ordered separately. Contact your Avantor representative for more information.
Customers Who Bought This Also Bought