Corning® Gosselin™ Transfer Pipettes, Corning
Supplier: Corning
Certificates
About this item
Corning® Gosselin™ offers a wide range of disposable sampling tools including Pipettes.
Use of disposable pipettes decreases cross-contamination risks. Polyethylene transfer pipettes are sterile and have metric graduations. Transfer pipettes are packaged in peelable bags.
Polypropylene straw pipettes are assured Sterile 10⁻³. They are packaged in tamper-evident zip bags.
Product traceability information is included on each bag.
Corning® Gosselin™ is a Corning brand of laboratory plasticware designed for use in industrial microbiology labs such as Food & Beverage, Environmental/Water Testing, etc.
See also Corning® Gosselin™ Dippers with Removable Handles, Cutlery, Blender Bags, and Inoculating Loops and Spreaders.
Customers Who Bought This Also Bought