Epredia™ Superfrost™ Excell™ Microscope Adhesion Slides, Thermo Scientific
Fournisseur: Erie Scientific
Certificats
A propos de cet article
Slide surface has superior adhesion and wettability
Slide surface has superior adhesion and wettability
Developed for use in HIER methods requiring high pH antigen retrieval solutions, including EDTA.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.