Pt 100 Precision Digital Thermometers, Thermco
Fournisseur: Thermco
Certificats
A propos de cet article
Precision RTD digital thermometers can be used as a temperature standard for the verification and calibration of laboratory equipment, as well as liquid-in-glass, digital, and dial bi-metal thermometers
Records minimum, maximum, and average temperatures. Devices feature a 7-segement LCD display and backlight. Waterproof
Certifications: Certified and traceable to NIST standards. Pt100 Platinum according to EN 60751.
Ordering information: Includes insertion probe and three AAA batteries. Other certified temperatures can be requested; please contact your Avantor representative for more information.
Spécifications
- Catalog No:
- 89137-110
- 10145-560
- 89137-112
- 10145-562
- 10145-558
- Temperature resolution:
- 0.1°C / °F
- 0.1°C / °F
- 0.1°C / °F
- 0.1°C / °F
- 0.1°C / °F
- Temperature accuracy:
- ±0.4°C (instrument only), ±1 digit
- ±0.4°C (instrument only), ±1 digit
- ±0.4°C (instrument only), ±1 digit
- ±0.4°C (instrument only), ±1 digit
- ±0.4°C (instrument only), ±1 digit
- W×D×H:
- 125×64×32 mm (6 × 2½ × 1¼”)
- 125×64×32 mm (6×2½×1¼”)
- 125×64×32 mm (6 × 2½ × 1¼”)
- 125×64×32 mm (6×2½×1¼”)
- 125×64×32 mm (6×2½×1¼”)