Hanging Dual Scale Thermometer, Thermco
# de catalogue 89168-870
Fournisseur: Thermco
Spécifications
- Cat. no.:89168-870
- Range:−40…+25 °C (−40…+80 °F)
- Supplier No.:BFZ80
Spécifications
A propos de cet article
This non-mercury hanging thermometer offers a dual scale of Celsius and Fahrenheit
It works for both refrigerators and freezers, and the tube is completely enclosed in plastic for safety. Includes stainless steel shelf hangers with keyhole backs for wall hanging
Customers Who Bought This Also Bought