Special Application Instrument Thermometers, Thermco®
Fournisseur: Thermco
Certificats
A propos de cet article
These non-mercury thermometers are calibrated against NIST standards to assure accuracy and reproducibility
Some thermometers have fluoropolymer coating. The certified thermometers include a deluxe, felt-lined storage box. Thermometer styles available include General-Purpose Certified Thermometers; Thermometers for Block Heaters; Thermometers for Blood Bank Refrigerators; and Thermometers for Water baths and Incubators.
Customers Who Bought This Also Bought