Vaccine Thermometer for Coolers, Thermco
# de catalogue 89168-868
Fournisseur: Thermco
Spécifications
- Ø:19 mm (¾”)
- Length:200 mm (8")
- Cat. no.:89168-868
- Divisions:0.5 °C
- Range:−30…+25 °C
- Supplier No.:ACC1482163
Spécifications
A propos de cet article
This precision refrigeration thermometer is encased in a water-tight, shock-proof plastic case and can easily be mounted in a vertical or horizontal orientation
The built-in cylindrical lens allows for an optically widended scale and easy reading
Customers Who Bought This Also Bought