Triple Display Certified Thermometers, Thermco
Fournisseur: Thermco
Certificats
A propos de cet article
These large digital thermometers simultaneously show the current, minimum, and maximum temperatures and update continuously
The probe is housed in an unbreakable plastic bottle filled with a buffering media that maintains the thermometer reading when the instrument door is opened. The thermometer and the probe bottle have dual magnets to attach to the instrument. Thermometers also feature a flip-open stand, wall mounting key hole, and an AAA alkaline battery.
An audible alarm sounds when the temperature rises above or below a set temperature. The minimum/maximum feature monitors critical temperature for any period of time
Certifications: Each thermometer is certified and traceable to NIST. Each thermometer is serialized.
Customers Who Bought This Also Bought