Shandon™ Neoprene Cork Dissecting Boards, Thermo Scientific
Fournisseur: Thermo Scientific Hamilton
Certificats
A propos de cet article
Work on a durable, cut-resistant surface over a marine plywood base with Shandon™ Neoprene Cork Dissecting Boards.
- Blended rubber and cork composition resists absorption and allows for thorough rinsing
- Rubber bumpers attached to each corner raise the cutting surface to 3.8 cm (1.5")
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.