A propos de cet article
This Logger is a battery powered stand alone device that records up to 64000 samples of temperature data.
- Push button or program to start recording
- Four digit LCD display
- Two user progammable alarms, high or low
- Indication of alarms, triggered, small compact size
- N.I.S.T. calibration available and MKT calculation included
- High accuracy and repeatability
The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery. The Track‐It™ Temperature Logger can be ordered with or without an LCD display and with a standard (one year) or long life battery (three year).
Information displayed on the LCD is also user programmable and can be sequenced by pressing the logger button. Both models have a multicolored LED indicator to notify user of alarms or general activity. A splash proof protective cover protects the logger when in use.
The unit is easily configured using the included Track‐It™ Software. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed.
Coin cell option: Battery: Lithium CR2032
Hi energy option: Battery: EF651625
Spécifications
- Catalog No:
- 89184-534
- 89184-532
- 89409-780
- 89409-778
- 89409-776
- 89184-530
- 89409-774
- 89184-528
- Interface:
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- W×D×H:
- 29.5×92.9×19.1 mm (1.16×3.66×0.75")
- 29.5×92.9×19.1 mm (1.16×3.66×0.75")
- 29.5×92.9×19.1 mm (1.16×3.66×0.75")
- 29.5×92.9×19.1 mm (1.16×3.66×0.75")
- 29.5×92.9×19.1 mm (1.16×3.66×0.75")
- 29.5×92.9×19.1 mm (1.16×3.66×0.75")
- 29.5×92.9×19.1 mm (1.16×3.66×0.75")
- 29.5×92.9×19.1 mm (1.16×3.66×0.75")