TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument | Avantor
TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument
Supplier:   Monarch Instrument
TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument
Supplier:  Monarch Instrument
Certificates
TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument
TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument
10 Options Available Below
Product Details & Documents
The pressure/temperature data logger is a battery powered stand alone water tight compact data logger that records up to 64000 samples of pressure and/or temperature data
The logger is packaged in a rugged submersible 316L stainless steel housing and has a standard ¹/₄" NPT fitting. The replaceable internal lithium battery has up to a 5 year life. The unit is easily configured using the included Track‐It™ software. Simply connect a standard USB cable to the logger and an open USB port on your PC and the Track‐It™ software automatically identifies the logger.
Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery.
It can be configured to record pressure and temperature or either parameter alone to maximize data storage space. The Track‐It™ pressure/temperature data logger can be ordered in four pressure ranges: 0 - 35, 0 - 150, 0 - 350 and 0 - 550 psi.
Certifications: CE marked.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89184-584
  • 89184-586
  • 89184-588
  • 89200-716
  • 89409-800
  • 89409-792
  • 89409-794
  • 89409-796
  • 89409-798
  • 89409-802
  • Alarm:
  • High/low alarm
  • High/low alarm
  • High/low alarm
  • High/low alarm
  • High/low alarm
  • High/low alarm
  • High/low alarm
  • High/low alarm
  • High/low alarm
  • High/low alarm
  • Interface:
  • USB
  • USB
  • USB
  • USB
  • USB
  • USB
  • USB
  • USB
  • USB
  • USB
  • Battery life:
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • 5 years (with 1 min sampling rate)
  • Data memory - No. of readings:
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • 32000/64000 (depends on setup)
  • Batteries:
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
  • 1/2×AA Lithium battery, 1.2 Ah
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Range
    Résolution
    Accuracy
    Availability
    Price
  • TRACK-IT™ 150 psi Pressure/Temperature Logger with long life battery
    P: 0 - 150 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,169.92
    $1,169.92
    Each
       $1,169.92
    Supplier Number: 5396-0301
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89184-584
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
    Small Business Enterprise
  • TRACK-IT™ 350 psi Pressure/Temperature Logger with long life battery
    P: 0 - 350 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,169.92
    $1,169.92
    Each
       $1,169.92
    Supplier Number: 5396-0302
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89184-586
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
    Small Business Enterprise
  • TRACK-IT™ 550 psi Pressure/Temperature Logger with long life battery
    P: 0 - 550 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,349.91
    $1,349.91
    Each
       $1,349.91
    Supplier Number: 5396-0303
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89184-588
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
    Small Business Enterprise
  • TRACK-IT™ 35 psi Pressure/Temperature Logger with long life battery
    P: 0 - 35 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,208.40
    $1,208.40
    Each
       $1,208.40
    Supplier Number: 5396-0304
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89200-716
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
    Small Business Enterprise
  • TRACK-IT™ 2000 psi Pressure/Temperature Logger with long life battery
    P: 0 - 2000 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,347.78
    $1,347.78
    Each
       $1,347.78
    Supplier Number: 5396-0305
    Spécifications:
    More
    Specifications
    Cat. No.
    89409-800
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
  • TRACK-IT™ 150 psi Pressure/Temperature Logger with long life battery, Calibrated
    P: 0 - 150 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,479.59
    $1,479.59
    Each
       $1,479.59
    Supplier Number: 5396-0301-CAL
    Spécifications:
    More
    Specifications
    Cat. No.
    89409-792
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
  • TRACK-IT™ 350 psi Pressure/Temperature Logger with long life battery, Calibrated
    P: 0 - 350 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,479.59
    $1,479.59
    Each
       $1,479.59
    Supplier Number: 5396-0302-CAL
    Spécifications:
    More
    Specifications
    Cat. No.
    89409-794
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
  • TRACK-IT™ 550 psi Pressure/Temperature Logger with long life battery, Calibrated
    P: 0 - 550 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,664.23
    $1,664.23
    Each
       $1,664.23
    Supplier Number: 5396-0303-CAL
    Spécifications:
    More
    Specifications
    Cat. No.
    89409-796
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
  • TRACK-IT™ 35 psi Pressure/Temperature Logger with long life battery, Calibrated
    P: 0 - 35 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,479.59
    $1,479.59
    Each
       $1,479.59
    Supplier Number: 5396-0304-CAL
    Spécifications:
    More
    Specifications
    Cat. No.
    89409-798
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
  • TRACK-IT™ 2000 psi Pressure/Temperature Logger with long life battery, Calibrated
    P: 0 - 2000 psi
    V: 0 - –380 torr; 0 - –760 torr
    T: –20…+85 °C
    T: 0.1 °C
    T: ±1 °C (0...50 °C), ±2 °C (−20...+85 °C)
    V: ±0.25%
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,664.23
    $1,664.23
    Each
       $1,664.23
    Supplier Number: 5396-0305-CAL
    Spécifications:
    More
    Specifications
    Cat. No.
    89409-802
    Alarm
    High/low alarm
    Interface
    USB
    Battery life
    5 years (with 1 min sampling rate)
    Data memory - No. of readings
    32000/64000 (depends on setup)
    Batteries
    1/2×AA Lithium battery, 1.2 Ah
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Battery: lithium 1/2AA-sized 1.2 AH
    Replacement Battery: lithium 1/2AA-sized 1.2 AH
    89184-602
    Log in pour la disponibilité
    Each - $80.18
Informations sur le produit
  • Catalog #
    Availability
    Price
  • TRACK-IT™ software replacement CD
    TRACK-IT™ software replacement CD
    89184-594
    Log in pour la disponibilité
    Each - $74.68
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Garden hose adaptor for Pressure logger
    Garden hose adaptor for Pressure logger
    89184-604
    Log in pour la disponibilité
    Each - $168.61
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacment O-rings for Pressure Logger- 3 pack
    Replacment O-rings for Pressure Logger- 3 pack
    89184-606
    Log in pour la disponibilité
    Pack of 3 - $29.09
Informations sur le produit
  • Catalog #
    Availability
    Price
  • 3 ft USB 2.0 interface cable
    3 ft USB 2.0 interface cable
    89184-592
    Log in pour la disponibilité
    Each - $29.86