A propos de cet article
The DC voltage and current data loggers are two channel battery powered stand alone compact data loggers that record up to 32000 samples each of DC voltage or current data
The unit is easily configured using the included Track‐It™ Software. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. After the logger has been programmed, disconnect it from the PC and attach the voltage or current module.
The DC voltage and current data loggers are two channel battery powered stand alone compact data loggers that record up to 32000 samples each of DC voltage or current data
The unit is easily configured using the included Track‐It™ Software. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. After the logger has been programmed, disconnect it from the PC and attach the voltage or current module.
The unit is easily configured using the included Track‐It™ Software. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. After the logger has been programmed, disconnect it from the PC and attach the voltage or current module.
The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery.
Units are available in 4 DC voltage ranges or for DC current. The loggers come with and LCD display with the option of a standard (1 year) or long life battery (3 year).
Spécifications
- Catalog No:
- 89184-552
- 89409-812
- 89409-810
- 89409-808
- 89409-806
- 89184-560
- 89409-804
- 89184-558
- 89184-556
- 89184-554
- 89184-566
- 89184-564
- 89184-562
- 89184-550
- 89184-548
- 89409-822
- 89409-820
- 89409-818
- 89409-816
- 89409-814
- Alarm:
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- High/low level alarms
- Display:
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- LCD, 4 digit
- Interface:
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- USB
- Battery life:
- 1 year (1 min sampling rate)
- 1 year (1 min sampling rate)
- 1 year (1 min sampling rate)
- 1 year (1 min sampling rate)
- 1 year (1 min sampling rate)
- 3 years (1 min sampling rate)
- 1 year (1 min sampling rate)
- 3 years (1 min sampling rate)
- 1 year (1 min sampling rate)
- 1 year (1 min sampling rate)
- 3 years (1 min sampling rate)
- 3 years (1 min sampling rate)
- 3 years (1 min sampling rate)
- 1 year (1 min sampling rate)
- 1 year (1 min sampling rate)
- 3 years (1 min sampling rate)
- 3 years (1 min sampling rate)
- 3 years (1 min sampling rate)
- 3 years (1 min sampling rate)
- 3 years (1 min sampling rate)
- W×D×H:
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- 25.4×103.2×16.7 mm
- Data memory - No. of readings:
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- 32000/64000 (depends on setup)
- Batteries:
- Lithium CR2032 battery
- Lithium CR2032 battery
- Lithium CR2032 battery
- Lithium CR2032 battery
- Lithium CR2032 battery
- Lithium EF65162 battery
- Lithium CR2032 battery
- Lithium EF65162 battery
- Lithium CR2032 battery
- Lithium CR2032 battery
- Lithium EF65162 battery
- Lithium EF65162 battery
- Lithium EF65162 battery
- Lithium CR2032 battery
- Lithium CR2032 battery
- Lithium EF65162 battery
- Lithium EF65162 battery
- Lithium EF65162 battery
- Lithium EF65162 battery
- Lithium EF65162 battery