Flexible Tip Digital Thermometer, Graham-Field
# de catalogue 95062-775
Fournisseur: GF HEALTH PRODUCTS INC. TE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Flexible Tip Digital Thermometer
- Supplier No.:L2214
Spécifications
A propos de cet article
Digital thermometer displays reading in Fahrenheit or Celsius within 10 seconds
Large digital display has memory recall and auto-off. Waterproof.
Customers Who Bought This Also Bought