Fabricel® Pillowcase, TIDI® | Avantor
Fabricel® Pillowcase, TIDI®
Full-Size Pillowcase, White, 53.3L x 76.2W cm (21 x 30")
Catalog # 97018-546
Supplier:   Tidi Products
Fournisseur #:  919355
Fabricel® Pillowcase, TIDI®
Catalog # 97018-546
Supplier:  Tidi Products
Fabricel® Pillowcase, TIDI®
Fabricel® Pillowcase, TIDI®
Full-Size Pillowcase, White, 53.3L x 76.2W cm (21 x 30")
Price
$332.16
Log in pour la disponibilité
Case of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Soft, tear-resistant pillowcase effectively combines barrier protection and patient comfort
Unique Fabricel® material fuses tissue and a layer of polyethylene film that helps prevent leak-through
Spécifications
  • Color:
    White
  • Désignation:
    Full-Size Pillowcase
  • Taille:
    53.3L x 76.2W cm (21 x 30")