Sterile Eye Pad, DUKAL™ Corporation
Fournisseur: Dukal
Certificats
A propos de cet article
Eye Pads are made of an absorbent cotton filled pad with a soft, gentle, non-woven cover that offers protection to this delicate area.
- Features an easy place tab on back to ensure clean placement
- Sealed edges provide minimal linting
- Not made with natural rubber latex
Packaging: Individually wrapped and packaged in a dispenser box for convenience
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.