Gauze Ball, DUKAL™ Corporation
Fournisseur: Dukal
Certificats
A propos de cet article
Sterile pressure dots.
- Non-woven gauze spheres that provide pressure to insertion site
- An excellent alternative to using a folded 4×4" gauze pad
- Not made with natural rubber latex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.