Alcohol Prep Pads, DUKAL™ Corporation
Supplier: Dukal
Certificates
About this item
These 2-ply alcohol prep pads are saturated with 70% isopropyl alcohol. Ideal for use as an antiseptic cleanser.
- Non-woven pads
- Not made with natural rubber latex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.