About this item
Now silent and effortless, Thermo Scientific™ F1-ClipTip™ Variable Volume Pipettes feature newly enhanced volume adjustment, and the added confidence of ClipTip technology that locks tips firmly to the pipette with a complete seal.
- Enhanced volume adjustment
- Complete seal
- Remains sealed until ejection
- High-quality results
- More efficient research
- Lightweight design
- Secure volume lock
With a light touch, each tip is clipped securely on the pipette. When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI. Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort. The 120° adjustable finger rest, for both right- and left-handed operator comfort, allows the hand to relax during pipetting cycles.
A 150% increase in air boost ensures efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below. Each tip is clipped securely on the pipette. Lifetime manufacturer warranty.