Corning® Reusable Plastic Single Metric Scale, Graduated Cylinder, PP, TC with Funnel Top, Corning
Supplier: Corning
Certificates
About this item
PP, translucent, graduated.
- Cylinders are manufactured with raised graduations at the primary scale points
- With hexagonal base, calibrated 'to contain' (TC)
- Exposure to temperatures up to 60 °C may not affect accuracy
Graduated cylinders Class B calibrated 'To Contain' (TC) according to DIN 12681/ISO 6706.
It is important to recognize the difference in specifications regarding DIN and ASTM standards. For reference, ASTM Class A and B specifications are more precise than DIN Class A and B specifications, respectively.
Specifications
- Catalog No:
- 89172-026
- 89172-028
- 89172-034
- 89172-036
- 89172-030
- 89172-032
- 89172-022
- 89172-024
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short