ContactUsLinkComponent
VWR Life Science Phenol-Free Total RNA Purification Kit
VWR Life Science Phenol-Free Total RNA Purification Kit
Catalog # 97064-308
Supplier:  VWR
   Safer Human and Environmental Health
CAS Number:  
KIT TOTAL RNA PURIFICATION PHENOL-FREE
VWR Life Science Phenol-Free Total RNA Purification Kit
Catalog # 97064-308
Supplier:  VWR
   Safer Human and Environmental Health
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Plant samples
    Bacteria
    Blood
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    PHENOL-FREE TOTAL RNA PURIFICATION KIT
  • Size:
    Kit
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Description:
    PHENOL-FREE TOTAL RNA PURIFICATION KIT
  • Cat. no.:
    97064-308
  • Supplier No.:
    N788-KIT
Specifications
About this item
VWR® Phenol-FreeTotal RNA Purification Kit provides a rapid method for the isolation and purification of total RNA from cultured animal cells, tissue samples, blood, bacteria, yeast, fungi, plants and viruses.
  • Save time
  • Reduce hazardous chemical use
  • Enhance recovery of total RNA including microRNA species
The kit purifies all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA). The RNA is preferentially purified from other cellular components, such as proteins, without the use of phenol or chloroform. The purified RNA is of the highest integrity, and can be used in a number of downstream applications including real time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays.
Ordering information: Lysis Solution, 40 ml
Wash Solution, 22ml
Elution Solution, 6 ml
Mini Spin Columns, 50
Collection Tubes, 50
Customers Who Bought This Also Bought