ContactUsLinkComponent
VWR® Agarose Gel Loading Dye, Ultra Pure Grade
VWR® Agarose Gel Loading Dye, Ultra Pure Grade
# de catalogue 97062-664
Fournisseur:  VWR
Numéro CAS:  
DYE AGRS GEL LOADING 6X ULT-PR GRD 5ML
VWR® Agarose Gel Loading Dye, Ultra Pure Grade
# de catalogue 97062-664
Fournisseur:  VWR
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    6X Gel Loading Dye
  • Size:
    5 ml (0.16 oz.)
  • Cat. no.:
    97062-664
  • Supplier No.:
    E190-5ML
Spécifications
A propos de cet article
VWR® electrophoresis gel loading dyes are tested for contaminating DNase, RNase, and protease activity and are sterile filtered for long shelf life and stability. In addition, all dyes are function tested and do not interfere with sample migration. Their ready-to-use format eliminates exposure to potentially irritating and harmful powdered dyes. Agarose Gel Loading Dye is designed to optimize loading nucleic acids in acrylamide and agarose gels by imparting color to the sample to ensure efficient sample distribution into each well. Its blend of three tracking dyes make estimating sample migration simple and reliable.
  • Ready-to-use
  • Eliminates exposure to irritating and harmful powdered dyes
  • DNase-, RNase-, and protease-free
  • Functionally and analytically tested
Dye #1 is a light blue dye that migrates at 4,000 base pairs in 1% agarose. Dye #2 is an indigo dye that migrates at 600 base pairs in 1% agarose. Dye #3 is a magenta dye that migrates at 10 base pairs in 1% agarose. The three dyes migrate independently from the samples, allowing estimation of the migration of the nucleic acids. Dyes contain 15% Ficoll and 30% glycerol.
Customers Who Bought This Also Bought