ContactUsLinkComponent
Bovine Rnase A (from Pancreas)
Bovine Rnase A (from Pancreas)
# de catalogue 97062-172
Fournisseur:  VWR
Numéro CAS:  
RNASE A,PANCREATIC 250MG
Bovine Rnase A (from Pancreas)
# de catalogue 97062-172
Fournisseur:  VWR
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Source:
    Pancreas
  • Species:
    Bovine
  • Size:
    250 mg
  • Form:
    Lyophilized powder
  • Enzyme name:
    Rnase A
  • Enzyme activity:
    ≥60 U/mg
  • Enzyme synonyms:
    Ribonuklease A
  • pH:
    pH 8.0
  • Supplier No.:
    0675-250MG
Spécifications
A propos de cet article
Ribonuclease A is an endoribonuclease that efficiently hydrolyzes RNA contaminants in DNA preparations from tissue or bacterial cell cultures. RNase A is used for a variety of molecular biology applications, most commonly for the hydrolysis of RNA that contaminates DNA preparations. This enzyme is used during plasmid purification without nicking or degrading plasmid.
  • Useful during DNA purification
  • High purity
  • Available in a ready-to-use solution
Customers Who Bought This Also Bought