ContactUsLinkComponent
VWR® SOB Broth, Biotechnology Grade
VWR® SOB Broth, Biotechnology Grade
# de catalogue 97063-454
Fournisseur:  VWR
Numéro CAS:  
MEDIUM SOB BROTH BIOTECH GRADE 500 G
VWR® SOB Broth, Biotechnology Grade
# de catalogue 97063-454
Fournisseur:  VWR
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    500 g
  • Cat. no.:
    97063-454
  • Application:
    Recombinant E. coli Strains Preparation
  • Supplier No.:
    J906-500G
Spécifications
A propos de cet article
SOB broth powder is used for the preparation of nutritionally rich growth medium and transformation of competent E
SOB Broth (Super Optimal Broth) growth medium is nutritionally rich, containing casein hydrolysate and yeast extract as sources of nitrogenous compounds and growth factors,and magnesium sulfate as a supply of magnesium ions necessary for DNA replication.
when dissolved at 25.5 g per liter of water,yields the formulation
Tryptone -------- 20 Grams/L
Yeast Extract- - -5 Grams/L
Sodium Chloride--0.5 Gram/L
Medium should be autoclaved or sterile-filtered. Allow the solution to cool to 50 °C or below before adding supplemental components and antibiotic. Add 20 mL of 1 M MgSO4.Store SOB Broth powder at room temperature (18 – 26 °C). Store prepared medium cold (2 – 8 °C).
coli.
Customers Who Bought This Also Bought