VWR® TRIS Glycine Buffer, 10X Liquid Concentrate
Fournisseur: VWR
Certificats
A propos de cet article
A 1X working solution contains 0.025M Tris base and 0.192 M Glycine.
- Convenient concentrations and packaging
pH (1X, Water) @25°C - 8.3 - 8.7
Customers Who Bought This Also Bought