VWR® Trypsin, Crystalline, Proteomics Grade
# de catalogue 97063-884
Fournisseur: VWR
Numéro CAS:
Spécifications
- Packaging:Amber Bottle
- Size:1 g
- Form:Crystalline
- Enzyme name:Trypsin
- Grade:Proteomics Grade
- Supplier No.:M150-1G
Spécifications
A propos de cet article
Trypsin, Crystalline, Proteomics Grade.
- EINECS: 232-650-8
- GHS Symbol 1: GHS08
- GHS Symbol 2: GHS07
- MDL: MFCD00082094
Customers Who Bought This Also Bought