About this item
Argos cell and tissue culture plates are optically clear for easy visualization of cells
Products are manufactured in a class 100,000 clean room environments under ISO 9001:9002 quality systems. All products are manufactured with 100% USP VI crystal clear virgin polystyrene and high quality PE to ensure the integrity of your cells. Our cell and tissue culture products are specially treated with patented vacuum-gas plasma machine that causes the extremely hydrophobic polystyrene surface to become negatively charged and hydrophilic. This allows the cells to attach more evenly, and consistent, giving you superior attachment and growth.
Convenient, peel-open, medical-style packaging; lot numbers are printed on the case for easy traceability. Sterilized by gamma radiation and certified non-pyrogenic.
Extremely smooth well area maximizes growth area; raised well rims of flat bottoms reduce evaporation. Alphanumeric notation on each well for easy identification. Plates have individual condensation rings and a one-way fit lid to reduce cross contamination. Plates are available in 6, 12, 24 and 96 wells and compatible with standard equipment.