Disposable Mini Plastic Serological Pipettes, Argos Technologies
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Argos brand mini pipettes were developed for use in confined spaces such as laminar hoods
Fiber free paper & plastic wrap ensures quick peel apart opening. Polystyrene, gamma sterilized & pyrogen free. Dispenser box allows for access of pipettes without opening top; color coded with universal stacking design.
These mini pipettes measure 9" and fit most serological pipettors. Smaller size reduces distance between sample and pipettor aiding in more ergonomic pipetting. Unique filter plug stays put and eliminates lint fibers. Pipettes feature color coded rings for ease in identification, easy-to-read graduations from +1 to 2%.
Customers Who Bought This Also Bought