A propos de cet article
AquaMax® Microplate Washers are configurable with user interchangeable heads for both 96- and 384-well microplate washing applications, including plate or cell washing.
- Interchangeable heads
- One-Tough AutoClean feature
- Includes 1 year parts and labor manufacturer's warranty
Wash heads can be changed in 30 seconds with no tools using the innovative head latch mechanism
AquaMax® Washers have several important maintenance features, since the lack of proper washer maintenance is the number one cause of washer failure. Using the One-Touch AutoClean allows the user to completely clean out the entire fluid path. Auto Shutdown leaves the system empty and dry after a user defined interval, preventing the buildup of contaminants.
No recalibration is necessary and all heads are recognized by the AquaMax® Washer base. By changing heads, users can effectively have 4 plate washers in the space of 1. The AquaMax® has an internal vacuum pump saving even more space on the benchtop and automatically preventing contamination.
Spécifications
- Catalog No:
- 97059-514
- CA89429-528
- CA89429-526
- 97059-518
- Wash times 384w:
- Single cycle, 100 µl/well, 12 sec; three cycles, 100 µl, 28 sec.
- Single cycle, 100 µl/well, 12 sec; three cycles, 100 µl, 28 sec.
- Single cycle, 100 µl/well, 12 sec; three cycles, 100 µl, 28 sec.
- Single cycle, 100 µl/well, 12 sec; three cycles, 100 µl, 28 sec.
- Wash times 96w:
- Single cycle, 300 µl/well, 11 sec; three cycles, 300 µl, 26 sec.
- Single cycle, 300 µl/well, 11 sec; three cycles, 300 µl, 26 sec.
- Single cycle, 300 µl/well, 11 sec; three cycles, 300 µl, 26 sec.
- Single cycle, 300 µl/well, 11 sec; three cycles, 300 µl, 26 sec.
- Dimensions:
- 21H×46.6W×51.7D cm
- 21H×46.6W×51.7D cm
- 21H×46.6W×51.7D cm
- 21H×46.6W×51.7D cm
- Dispensing volume:
- 10 - 350 µl/well
- 10 - 350 µl/well
- 10 - 350 µl/well
- 10 - 350 µl/well
- Fast wash times (optimised for speed):
- 96-well wash head, 300 µl/well, three cycles, 14 sec; 384-well wash head, 100 µl/well, three cycles, 16 sec
- 96-well wash head, 300 µl/well, three cycles, 14 sec; 384-well wash head, 100 µl/well, three cycles, 16 sec
- 96-well wash head, 300 µl/well, three cycles, 14 sec; 384-well wash head, 100 µl/well, three cycles, 16 sec
- 96-well wash head, 300 µl/well, three cycles, 14 sec; 384-well wash head, 100 µl/well, three cycles, 16 sec
- Precision:
- 96-well: < or = 4% CV at 300 µl; 384-well: < or = 3% CV at 100 µl
- 96-well: < or = 4% CV at 300 µl; 384-well: < or = 3% CV at 100 µl
- 96-well: < or = 4% CV at 300 µl; 384-well: < or = 3% CV at 100 µl
- 96-well: < or = 4% CV at 300 µl; 384-well: < or = 3% CV at 100 µl
- Shaking speed:
- 3 Speeds up to 600 sec. max.
- 3 Speeds up to 600 sec. max.
- 3 Speeds up to 600 sec. max.
- 3 Speeds up to 600 sec. max.