ContactUsLinkComponent
Hematology Analyzer Slide, Azer Scientific
Hematology Analyzer Slide, Azer Scientific
# de catalogue 89178-326
Fournisseur:  Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
SLIDE HEMATOLOGY ANLYZR CS1440
Hematology Analyzer Slide, Azer Scientific
# de catalogue 89178-326
Fournisseur:  Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Hematology analyzer slide
  • Supplier No.:
    EMS100TPB
Spécifications
A propos de cet article
Azer Scientific's Hematology Analyzer Slides are designed for use in automated hematology slide makers.
  • Pure white glass
  • Two white, printed bars to prevent jamming
  • Beckman Coulter® systems compatible
  • 75 x 25 x 1 mm
  • Clipped corners
Manufactured from 100% pure white glass for the highest quality in hematology applications, these slides also feature two printed bars to help prevent jamming in equipment.
They are compatible with Beckman Coulter® systems.
Packaging: 72 per box, 20 boxes per case
Customers Who Bought This Also Bought