VWR® Complete Fly Workstation
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
Complete Fly Workstation includes FlyBed, FlowMate, FlyShot, tubing, connectors, and two FlyBed replacement pads.
- Built-in connectors for ease of use
- Equipped to handle FlyBed panel and FlyShot blow gun
FlowMate CO2 flow regulator aids in the anesthetizing of flies
FlyBed is used as a platform for examining anesthetized drosophila. Once anesthetized, drosophila are placed on the pad for easy viewing under a microscope. Specially designed edge allows for easy placement into fly vials. The panel is equipped with small openings to aid in the diffusion of CO2.
Customers Who Bought This Also Bought