About Face™ Cleansing Wipes, Micronova
Catalog # 89140-690
Supplier: Micronova
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:About Face™ cleansing wipe
- Material:Polyblend fabric
- Cat. no.:89140-690
- Packaging:Individual packets
- Supplier No.:MMR-01
Specifications
About this item
A pre-moistened cleansing wipe that is gentle enough for the face and that can be used to remove surface soil prior to gloving or sanitizing hands.
- Individually packaged, polyblend cleansing wipe
- Pre-moistened with a gentle, alcohol-free cleansing lotion
- Highly effective as a make-up remover
- Removes surface soil prior to gloving or sanitizing hands
About Face™ Cleansing Wipes are individually packaged and ideal for pre-gowning rooms.
Customers Who Bought This Also Bought