Line Valve, Ball, Stainless Steel, Rotarex
# de catalogue 89130-356
Fournisseur: Ceodeux Inc.
Spécifications
- Description:Ball line valve
- Cat. no.:89130-356
- Supplier No.:4SBV2TSS
Spécifications
A propos de cet article
Used for instrumentation, hydraulic pipes, pneumatic pipes, chromatography, control panels, moderate, vacuum systems, chemical research, gas analysis, etc.
Diaphragm sealing.
The assembly is made quickly by inserting the tube in the ready-to-use fitting and tighten. They require no special tools.
Ordering information: Additional materials of construction and end connections are available available; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought