Airguard® Activator™ Self-Sealing Panel Air Filters, Total Filtration
Fournisseur: Total Filtration Services
Certificats
A propos de cet article
Activator™ series panels and links consist of layers of multi-denier polyester media sealed around a heavy-duty 9-gauge internal wire frame with integral cross wire support members.
Applications include general ventilation, pre-filters, stand-alone single-stage filtration, built-up frame banks, side access systems, rooftop air handlers, air supply for paint spray booths, and economical upgrade from fiberglass or synthetic-framed disposable filters.
Customers Who Bought This Also Bought