GE Water™ Hytrex® Depth Filter Cartridges, Total Filtration
Fournisseur: Total Filtration Services
Certificats
A propos de cet article
High-performance filters combine efficiency, long-life, and purity.
Applications include high-purity chemicals, bottled water, pre-treatment for reverse osmosis, oil and gas, and electronics.
The purity and reliability of these cartridge filters ensure consistent results. Thermally bonded microfibers create a strong secure cartridge that traps particles throughout its depth.
Customers Who Bought This Also Bought