AirguardĀ® Synthetic Pocket Filters, Total Filtration
Fournisseur: Total Filtration Services
Certificats
A propos de cet article
Extended surface pocket filters feature efficiency, high dust holding capacity, long service life, and low resistance to air flow.
Applications include general ventilation, stand-alone single-stage filtration, built-up frame banks, side-access systems, rooftop air handlers, air supply for paint spray booths, and economical upgrade from fiberglass or synthetic-framed disposable filters.
Ordering information: Extended surface pocket filters are available in ratings from MERV 9 to MERV 14; contact your Avantor representative for more information.
SpƩcifications
- Catalog No:
- 78001-462
- 78001-530
- 78001-460
- 78001-538
- 78001-534
- 78001-544
- 78001-546
- 78001-540
- 78001-548
- 78001-470
- 78001-476
- 78001-478
- 78001-472
- 78001-550
- 78001-552
- 78001-480
- 78001-482
- 78001-558
- 78001-556
- 78001-564
- 78001-498
- 78001-496
- 78001-506
- 78001-574
- 78001-508
- 78001-504
- 78001-570
- 78001-580
- 78001-576
- 78001-440
- 78001-438
- 78001-448
- 78001-510
- 78001-444
- 78001-512
- 78001-524
- 78001-520
- 78001-526
- 78001-528
- 78001-454
- 78001-456
- 78001-450
- 78001-452
- Dimensions:
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 61 cm (24 x 24")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 61 x 30.5 cm (24 x 12")
- 30.5 x 61 cm (12 x 24")
- 61 x 51 cm (24 x 20")
- 61 x 51 cm (24 x 20")
- 51 x 51 cm (20 x 20")
- 61 x 51 cm (24 x 20")
- 51 x 51 cm (20 x 20")
- 51 x 61 cm (20 x 24")
- 61 x 51 cm (24 x 20")
- 51 x 51 cm (20 x 20")
- 61 x 51 cm (24 x 20")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
- 61 x 61 cm (24 x 24")
SpƩcifications
Customers Who Bought This Also Bought