Airguard® DP® Pleated Filters, MERV 8, Total Filtration
Fournisseur: Total Filtration Services
Certificats
A propos de cet article
DP® filters are made of 100% synthetic fibers that provide mechanical efficiency for a MERV 8 performance.
Applications include general ventilation, pre-filters, stand-alone single-stage filtration, built-up frame banks, side access systems, rooftop air handlers, air supply for paint spray booths, and economical upgrade from fiberglass or synthetic-framed disposable filters.
Customers Who Bought This Also Bought