A propos de cet article
A variety of traditional wet mops for production facility and controlled environment cleaning
- C-Pro mops deliver both a high absorbency and superior durability
- MicroEco® mop is made from recycled PET bottles
- Sentrex mops are ideal for rugged cleaning on rough surfaces and high traffic areas
- Winger™ is ideal for all production facilities
- Looped-end construction allows for maximum floor coverage while eliminating tangling, fraying, and linting
- Microfiber technology minimizes linting
- Aluminum telescopic handle is lightweight and strong, and can be used with all mop sizes
The wet mop line includes a blue poly-coated mesh headband for maximum performance and durability manufactured with quality stitching. Tailband tape prevents tangling, provides wider surface coverage and improves commercial launderability. The wet mop line addresses key concerns with traditional wet mopping, absorbency, durability and performance. Quality synthetic yarns provide higher absorbency allowing faster drying times and greater surface area to be cleaned, as well as durability increasing the longevity of the mops. A high degree of debris pick-up on a wide variety of floor surfaces provides excellent performance. C-Pro cotton cut end narrow band wet mops are ideal for rugged cleaning in high traffic areas. These mops deliver both a high absorbency and superior durability. All mops are individually poly-bagged. Constructed of specially woven strands of low-linting polyester/cotton, the Ruff-T mop absorbs twice as much liquid as conventional mops making it ideal for non-slip, non-skid surfaces. The economical MicroEco® looped-end wide band wet mop features a unique four-ply yarn manufactured from a combination of microfiber yarn and green PET synthetic fibers derived from post-consumer plastic bottles. The advantages of Microfiber yarn include minimized linting and high resistance to chemicals and disinfectants. Six-row stitched. Looped-end construction allows for maximum floor coverage while eliminating tangling and fraying. Mop is individually poly-bagged.
The Ruff-T looped mop stands up to rough floor surfaces such as stone, terrazzo, concrete, and other textured surfaces. This low-linting highly absorbent mop is ideal for controlled environments, healthcare, and general production facilities. Mop is autoclaveable and poly-bagged. Sentrex cut end narrow band wet mop is ideal for rugged cleaning on rough surfaces and high traffic areas. Made from a highly durable synthetic/rayon blended yarn with superior tensile strength properties, it can be used for floor finishing and waxing. Delivers high absorbency and can withstand repeated laundering without linting. Heavy-duty poly-coated mesh headband withstands harsh chemicals, launderings for the life of the mop, and abuse when the headband is used as a scrubber. Mop is individually poly-bagged.
Sentrex Looper looped-end wide band wet mop is ideal for rugged cleaning on rough surfaces and high traffic areas. Made from a highly durable synthetic/rayon blended yarn with superior tensile strength properties. Delivers high absorbency and can withstand repeated laundering without linting. Heavy-duty poly coated mesh headband can withstand harsh chemicals, laundering for the life of the mop, and abuse when headband is used as a scrubber. Looped-end construction allows for maximum floor coverage while eliminating tangling and fraying. Mop is individually poly-bagged and available in white. Winger® looped-end wide band wet mop is ideal for all production facilities. Superior strength and absorbency. No cotton used. Looped-end construction allows for maximum floor coverage while eliminating tangling, fraying, and linting. Use blue mops in coordination with white mops to avoid cross-contamination. Mop is individually poly-bagged and available in blue.
Mop stick features a wood handle. Twist action secures mop head onto stick. Clamp opens for easy installation. Mop stick can be used with all mop sizes.
Snap-N-Go® telescopic handle is constructed of lightweight but strong aluminum. Ergonomic center grip and anti-slip handle grip. Changeable color coding clips allow for coding of mops (blue, red, yellow, green). Mop handle can be used with all mop sizes.