ContactUsLinkComponent
The Looper Looped-End Wet Mop, Wide Band, Vileda Professional-FHP
The Looper Looped-End Wet Mop, Wide Band, Vileda Professional-FHP
# de catalogue 89134-278
Marino Looper White Looped-End Mops
The Looper Looped-End Wet Mop, Wide Band, Vileda Professional-FHP
# de catalogue 89134-278
Spécifications
  • Description:
    Mop Head, Blue
  • Cat. no.:
    89134-278
  • Length:
  • Supplier No.:
    131598
  • Headband Size:
    5"
Spécifications
A propos de cet article
Heavy-duty poly coated headband mesh withstands harsh chemicals, and abuse when headband is used a scrubber.
  • The Looper is ideal for hospital or nursing home facilities
  • Superior strength and absorbency; no cotton used
  • Looper-end construction allows for maximum floor coverage while eliminating tangling, fraying, and linting
Professional janitor wood mop stick features a clamp that opens for easy installation. Twist action secures mop head. For use with all size mop heads.
Strong, lightweight Snap-N-Go™ telescopic handle is constructed of aluminum with an ergonomic center grip and anti-slip handle grip. Changeable color coding clips allow for coding of mops (blue, red, yellow, green).
Made from a unique blend of yarn that enables superior absorbency while withstanding repeated laundering and eliminating linting. No cotton used. Looped-end construction and fan-tail design allows for maximum floor coverage while eliminating linting, tangling, and fraying. Six-row stitched.
Ordering information: Mop head and handle are sold separately. Mop heads are individually poly-bagged.
Customers Who Bought This Also Bought