About this item
Designed for all critical cleanroom surfaces, PharmaMOP® Mop System allows cleaning of ceilings, walls, and floors with ease.
- Mop heads feature polyester over foam construction
- Abrasion-free mopping for ISO Class 1–7 cleanrooms
- Gamma irradiated to SAL 10¯⁶
System combines a unique mop head optimized to meet label-indicated disinfectant contact times with an ultra-lightweight, telescoping handle and frame for unsurpassed cleaning efficiency
Mop frame features a two-direction 360° swivel and 3.8cm (1.5") low-profile design that enables unsurpassed access to tight spaces, reducing uncleanable dead spots. Adjustable to lengths of 127cm (50"), 144.8cm (57"), and 162.6cm (64").
Enhances SOP cleaning compliance through improved ergonomics. Utilizes a simple press fit connection that enables gloved operators to easily attach mop heads. Compatible with steam sterilization processes.