UltraMOP™ Mop Head for Cleanroom Environments, Foamtec International
Fournisseur: Foamtec International
CE Compliant
Certificats
A propos de cet article
Perfex®-style cleanroom foam mop head is designed to fit the TruCLEAN® mopping system
Mop head is constructed of a snag-resistant, polyester fabric lamination that extends over the leading and trailing edges of the underlying foam sponge, eliminating the need for separate polyester socks. This unique Bump-PROOF® design improves cleanliness and durability. Compatible with Gamma, ETO, and autoclave sterilization
Certifications: Lot certified, individually packed, and lot traceable back to raw materials.
Ordering information: Handle and frame not included. Fits Perfex-style mop frame nos. 2238 and 2239 (see 22940-020 series). Foam mop refill is individually packed to maintain cleanliness.
Customers Who Bought This Also Bought