VWR® Irradiated Non-Skid High-Top Boot Cover
Catalog # 414004-501
Supplier: VWR International
Specifications
- Shoe size:XL
- Garment application:Cleanroom
- Color:White
- Height:45.7 cm (18")
- Length:38.1 cm (15")
- Bacterial filtration efficiency:>99.9%
- Particle shedding (Helmke Drum):Level I
- Water resistance:Pass
- Tensile strength:29.4 (Filling)
- Tearing strength:5.7 (Widthwise Yarns)
- Synth. blood penetration resistance:Pass
- Thickness:0.22 mm
- Weight:63 g/m²
- Cat. no.:414004-501
Specifications
About this item
Irradiated non-skid high-top boot covers have been engineered to optimize skid resistance, barrier protection, cleanliness, and comfort.
- Non-skid soles provide maximum traction on wet and smooth surfaces
- Superior coefficient of friction
- Excellent fluid and particulate barrier
- Soft, cloth-like fabric with maximum tear strength
Boot covers are gamma irradiated to a Sterility Assurance Level (SAL) of 10-4 to address the concern of bio-burden contamination. Manufactured in an ISO Class 8 (FED-STD-209E Class 100,000/M6.5) cleanroom.
Certifications: Boot covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Ordering information: Bulk packed in sealed polyethylene bags contained within a larger carton liner. Each case is supplied with a certificate of irradiation.
Customers Who Bought This Also Bought