Trigger Sprayer for Accel® TB Disinfectant, Contec®
Trigger Sprayer for Accel® TB Disinfectant, Contec®
Trigger Sprayer for 946.3 ml (32 oz.) Bottle
Catalog # 89066-160
Supplier:   CONTEC, INC. MX
Supplier #:  ACCTRIGR
Trigger Sprayer for Accel® TB Disinfectant, Contec®
Catalog # 89066-160
Supplier:  CONTEC, INC. MX
Trigger Sprayer for Accel® TB Disinfectant, Contec®
Trigger Sprayer for Accel® TB Disinfectant, Contec®
Trigger Sprayer for 946.3 ml (32 oz.) Bottle
Price
$105.78
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 12 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Non-aerosol trigger sprayers for 32 oz. bottle.
Spécifications
  • Désignation:
    Trigger Sprayer for 946.3 ml (32 oz.) Bottle