CONSTIX®, Sealed Foam Swabs, SF-2 Series, Contec®
# de catalogue 68310-230
Fournisseur: Contec
Small Business Enterprise
Spécifications
- Description:Sealed Foam Swabs, SF-2 Series
- Cat. no.:68310-230
- Handle Length:101.6 mm (4")
- Overall Length:127 mm (5")
- Head Width:15.7 mm (0.62")
- Head Length:25.4 mm (1")
- Handle Width:6.5 mm (0.26")
- Supplier No.:SF-2
Spécifications
A propos de cet article
CONSTIX® SF-2 swabs consist of 100ppi sealed closed-cell foam head with a polypropylene handle. The swab head is thermally sealed and bonded to the handle without adhesives or binders.
- Large rectangular head ideal for swabbing larger areas and applying coatings
The polyurethane foam head provides good particle entrapment and removal. The large size of the foam head of the SF-2 swab provides better surface coverage for cleaning. The SF-2 swab is compatible with many solvents, such as isopropanol and ethanol, but is not recommended for use with ketones, such as acetone or MEK.
Ordering information: Laundered and double bagged.
Customers Who Bought This Also Bought