ContactUsLinkComponent
CONSTIX®, Sealed Foam Swabs, SF-1 Series, Contec®
CONSTIX®, Sealed Foam Swabs, SF-1 Series, Contec®
Catalog # 68310-228
Supplier:  Contec
Small Business Enterprise
undefined
CONSTIX®, Sealed Foam Swabs, SF-1 Series, Contec®
Catalog # 68310-228
Supplier:  Contec
Supplier Number:  SF-1
Small Business Enterprise
Specifications
  • Description:
    Sealed Foam Swabs, SF-1 Series
  • Cat. no.:
    68310-228
  • Handle Length:
    102.6 mm (4.04")
  • Overall Length:
    127 mm (5")
  • Head Width:
    20.3 mm (0.8")
  • Head Length:
    24.4 mm (0.96")
  • Handle Width:
    6.5 mm (0.26")
  • Supplier No.:
    SF-1
Specifications
About this item
CONSTIX® SF-1 sealed foam swabs consist of polyurethane foam bonded to a polypropylene handle without the use of adhesives or binders.
  • Polyurethane foam head provides good particle entrapment and removal
  • Large head is ideal for cleaning and applying coating
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. Though compatible with many solvents, the SF-1 is not recommended for use with ketones, such as acetone or MEK.
Ordering information: Laundered and double bagged.