FILTRA-BAG® Blender Bags, Labplas
Fournisseur: Labplas
Certificats
A propos de cet article
FILTRA-BAG ® blender bags are designed to simplify taking an aliquot when working with samples which contain large amounts of residue and/or semi-solid / solid substances.
When lacing an aliquot onto growth media, it is very important that the volume of the aliquot be accurate and that it be free of solid particles. This prevents pipette blockage as well as uneven plate distribution.
Customers Who Bought This Also Bought