A propos de cet article
Denatured ethanol formulated to 70% with a small percentage of methyl alcohol and 30% USP Water for Injection which can be used for the disinfection and decontamination of cleanrooms, controlled areas, hard non-porous environmental surfaces, and many other applications that require the use of a sterile alcohol solution.
- Filled in ISO 5 (Grade A/B, Former Class 100)
- Prefiltered to 0.2 um
- Completely traceable and validated for sterility and shelf life
- Gamma irradiated to a sterility assurance level of 10^-6.
- Assayed according to current USP compendium
Applications include: Non-food contact, hard non-porous and inanimate surfaces, machinery, tools, tables, counters, laminar flow benches, floors, walls and carts. Also for shelves made of hard non-porous materials, such as glass, plastic, vinyl, chrome, and stainless steel.