VWR® Reclosable Bags with Writing Area
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
Reclosable bags are full gauge and offer exceptional clarity and quality.
- Reclosable design
- White writing area is compatible with markers or pens
- Reinforced zipper locks in a smooth motion
Red line bags have a red line along the top of the bag. Made of 100% virgin Low Density Polyethylene (LDPE) resin.
Certifications: Meet FDA/USDA specifications.
Customers Who Bought This Also Bought