VWR® Gusseted Poly Bags on Rolls
Supplier: VWR International
Certificates
About this item
VWR® clear Gusseted bags on a roll are made of 100% virgin Low Density Polyethylene (LDPE) resin.
- Excellent vapor and moisture barrier
- Can be used as a liber or pallet cover
- Three times the impact resistance and twice the tensile strength of traditional LDPE bags
Easy dispensing with perforations between bags. Tubular construction throughout with a durable bottom seal, make these bags the best on the market. This premium bag features superior clarity for showing the quality of your customers products. Wound on a 3" core with 1" Ø core plug. Can be heat sealed, tied, stapled or taped shut.
Certifications: Meet FDA/USDA specifications.
Customers Who Bought This Also Bought